Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 14 - 23417783 - 23417783 [+] on 23417783 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-208b-3p | MIMAT0004960 |
| Human | hsa-miR-208b-5p | MIMAT0026722 |
Stem-loop Details
Human

Genomic Map