Assay Details
Mature miRNA Sequence: | CGGGGCCGGGGCGCUGUCUGAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gadus morhua | gmo-miR-128-2-5p | MIMAT0044085 |
| Salmo salar | ssa-miR-128-3-5p | MIMAT0032316 |
Stem-loop Details
Gadus morhua
Salmo salar

Genomic Map