Assay Details
Mature miRNA Sequence: | CGGGGCCGGGGCGCUGUCUGAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gadus morhua | gmo-miR-128-2-5p | MIMAT0044085 |
| Salmo salar | ssa-miR-128-3-5p | MIMAT0032316 |
Stem-loop Details
Gadus morhua
Salmo salar
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gmo-miR-128-2-5p |
|---|---|
| miRBase Accession Number: | MI0036028 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGGGCCGGGGCGCUGUCUGAGA |
| Species: | Gadus morhua, Salmo salar |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | CGGGGCCGGGGCGCUGUCUGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gadus morhua | gmo-miR-128-2-5p | MIMAT0044085 |
| Salmo salar | ssa-miR-128-3-5p | MIMAT0032316 |