Assay Details
Mature miRNA Sequence: | AUGCACCUGGGCAAGGAUUCUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Callithrix jacchus | cja-miR-502b | MIMAT0049303 | |
| Daubentonia madagascariensis | dma-miR-502b | MIMAT0049269 | |
| Gorilla gorilla | ggo-miR-502b | MIMAT0024308 | |
| Microcebus murinus | mmr-miR-502b | MIMAT0049183 | |
| Nomascus leucogenys | nle-miR-502b | MIMAT0049363 | |
| Otolemur garnettii | oga-miR-500 | MIMAT0049690 | |
| View More Rows | |||
| Otolemur garnettii | oga-miR-502b | MIMAT0049691 | |
| Papio hamadryas | pha-miR-502b | MIMAT0049564 | |
| Rhesus monkey | mml-miR-500a-3p | MIMAT0026888 | |
Stem-loop Details
Callithrix jacchus
Daubentonia madagascariensis
Gorilla gorilla
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Rhesus monkey

Genomic Map