Assay Details
Mature miRNA Sequence: | AUGCACCUGGGCAAGGAUUCUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Callithrix jacchus | cja-miR-502b | MIMAT0049303 |
| Daubentonia madagascariensis | dma-miR-502b | MIMAT0049269 |
| Gorilla gorilla | ggo-miR-502b | MIMAT0024308 |
| Microcebus murinus | mmr-miR-502b | MIMAT0049183 |
| Nomascus leucogenys | nle-miR-502b | MIMAT0049363 |
| Otolemur garnettii | oga-miR-500 | MIMAT0049690 |
| View More Rows | ||
| Otolemur garnettii | oga-miR-502b | MIMAT0049691 |
| Papio hamadryas | pha-miR-502b | MIMAT0049564 |
| Rhesus monkey | mml-miR-500a-3p | MIMAT0026888 |
Stem-loop Details
Callithrix jacchus
Daubentonia madagascariensis
Gorilla gorilla
Microcebus murinus
Nomascus leucogenys
Otolemur garnettii
Papio hamadryas
Rhesus monkey

Genomic Map