Assay Details
Mature miRNA Sequence: | UGACAUCAUAUGUACGGCUGCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Alligator mississippiensis | ami-miR-489-3p | MIMAT0038296 | |
Astatotilapia burtoni | abu-miR-489 | MIMAT0042149 | |
Columba livia | cli-miR-489-3p | MIMAT0038657 | |
Cyprinus carpio | ccr-miR-489 | MIMAT0026308 | |
Gadus morhua | gmo-miR-489-3p | MIMAT0044024 | |
Ictalurus punctatus | ipu-miR-489 | MIMAT0029520 | |
View More Rows | |||
Metriaclima zebra | mze-miR-489 | MIMAT0042373 | |
Neolamprologus brichardi | nbr-miR-489 | MIMAT0042557 | |
Oreochromis niloticus | oni-miR-489 | MIMAT0042732 | |
Pundamilia nyererei | pny-miR-489 | MIMAT0042939 | |
Salmo salar | ssa-miR-489-3p | MIMAT0032658 |
Stem-loop Details
Alligator mississippiensis
Astatotilapia burtoni
Columba livia
Cyprinus carpio
Gadus morhua
Ictalurus punctatus
Metriaclima zebra
Neolamprologus brichardi
Oreochromis niloticus
Pundamilia nyererei
Salmo salar
