Assay Details
Mature miRNA Sequence: | UGACAUCAUAUGUACGGCUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-489-3p | MIMAT0038296 |
| Astatotilapia burtoni | abu-miR-489 | MIMAT0042149 |
| Columba livia | cli-miR-489-3p | MIMAT0038657 |
| Cyprinus carpio | ccr-miR-489 | MIMAT0026308 |
| Gadus morhua | gmo-miR-489-3p | MIMAT0044024 |
| Ictalurus punctatus | ipu-miR-489 | MIMAT0029520 |
| View More Rows | ||
| Metriaclima zebra | mze-miR-489 | MIMAT0042373 |
| Neolamprologus brichardi | nbr-miR-489 | MIMAT0042557 |
| Oreochromis niloticus | oni-miR-489 | MIMAT0042732 |
| Pundamilia nyererei | pny-miR-489 | MIMAT0042939 |
| Salmo salar | ssa-miR-489-3p | MIMAT0032658 |
Stem-loop Details
Alligator mississippiensis
Astatotilapia burtoni
Columba livia
Cyprinus carpio
Gadus morhua
Ictalurus punctatus
Metriaclima zebra
Neolamprologus brichardi
Oreochromis niloticus
Pundamilia nyererei
Salmo salar

Genomic Map