Assay Details
Mature miRNA Sequence: | UCCUCAUUGUGCAUGCUGUGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Astatotilapia burtoni | abu-miR-730 | MIMAT0042159 | |
| Cyprinus carpio | ccr-miR-730 | MIMAT0026317 | |
| Ictalurus punctatus | ipu-miR-730 | MIMAT0029524 | |
| Metriaclima zebra | mze-miR-730 | MIMAT0042386 | |
| Neolamprologus brichardi | nbr-miR-730 | MIMAT0042570 | |
| Oreochromis niloticus | oni-miR-730 | MIMAT0042755 | |
| View More Rows | |||
| Pundamilia nyererei | pny-miR-730 | MIMAT0042952 | |
| Salmo salar | ssa-miR-730a-5p | MIMAT0032679 | |
Stem-loop Details
Astatotilapia burtoni
Cyprinus carpio
Ictalurus punctatus
Metriaclima zebra
Neolamprologus brichardi
Oreochromis niloticus
Pundamilia nyererei
Salmo salar

Genomic Map