Assay Details
Mature miRNA Sequence: | UCCUCAUUGUGCAUGCUGUGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Astatotilapia burtoni | abu-miR-730 | MIMAT0042159 |
| Cyprinus carpio | ccr-miR-730 | MIMAT0026317 |
| Ictalurus punctatus | ipu-miR-730 | MIMAT0029524 |
| Metriaclima zebra | mze-miR-730 | MIMAT0042386 |
| Neolamprologus brichardi | nbr-miR-730 | MIMAT0042570 |
| Oreochromis niloticus | oni-miR-730 | MIMAT0042755 |
| View More Rows | ||
| Pundamilia nyererei | pny-miR-730 | MIMAT0042952 |
| Salmo salar | ssa-miR-730a-5p | MIMAT0032679 |
Stem-loop Details
Astatotilapia burtoni
Cyprinus carpio
Ictalurus punctatus
Metriaclima zebra
Neolamprologus brichardi
Oreochromis niloticus
Pundamilia nyererei
Salmo salar

Genomic Map