Assay Details
Gene Family ID: | MIPF0000445, mir-1225 |
Mature miRNA Sequence: | UGAGCCCCUGUGCCGCCCCCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1225-3p | MIMAT0005573 |
| Pongo pygmaeus | ppy-miR-1225-3p | MIMAT0016161 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map