Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UCGACAGCACGACACUGCCUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-196b-3p | MIMAT0009201 |
Mouse | mmu-miR-196b-3p | MIMAT0017170 |
Pteropus alecto | pal-miR-196b-3p | MIMAT0040010 |
Stem-loop Details
Human
Mouse
Pteropus alecto
