Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 7 - 27169783 - 27169783 [-] on 27169783 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-196b-5p | MIMAT0001080 |
Human | hsa-miR-196b-3p | MIMAT0009201 |
Stem-loop Details
Human
