Assay Details
Gene Family ID: | MIPF0000938, mir-2355 |
Mature miRNA Sequence: | AUUGUCCUUGCUGUUUGGAGAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-2355-3p | MIMAT0017950 |
| Bovine | bta-miR-2355-3p | MIMAT0011892 |
| Callithrix jacchus | cja-miR-2355 | MIMAT0039387 |
Stem-loop Details
Human
Bovine
Callithrix jacchus

Genomic Map