Assay Details
Amplicon Length: | 120 |
Chromosome Location: | Chr. 2 - 207109837 - 207109837 [+] on 207109837 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-2355-5p | MIMAT0016895 |
Human | hsa-miR-2355-3p | MIMAT0017950 |
Stem-loop Details
Human
