Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AUAAUACAUGGUUAACCUCUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-655-3p | MIMAT0003331 |
| Callithrix jacchus | cja-miR-655 | MIMAT0039376 |
| Guinea pig | cpo-miR-655-3p | MIMAT0047362 |
| Horse | eca-miR-655 | MIMAT0013171 |
| Pongo pygmaeus | ppy-miR-655 | MIMAT0016089 |
| Rabbit | ocu-miR-655-3p | MIMAT0048548 |
Stem-loop Details
Human
Callithrix jacchus
Guinea pig
Horse
Pongo pygmaeus
Rabbit

Genomic Map