Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 14 - 101049249 - 101049249 [+] on 101049249 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-655-5p | MIMAT0026626 |
| Human | hsa-miR-655-3p | MIMAT0003331 |
Stem-loop Details
Human

Genomic Map