Assay Details
Gene Family ID: | MIPF0000431, mir-744 |
Mature miRNA Sequence: | UGCGGGGCUAGGGCUAACAGCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-744-5p | MIMAT0004945 |
| Mouse | mmu-miR-744-5p | MIMAT0004187 |
| Bovine | bta-miR-744 | MIMAT0009369 |
| Dasypus novemcinctus | dno-miR-744-5p | MIMAT0048008 |
| Guinea pig | cpo-miR-744-5p | MIMAT0047379 |
| Pan troglodytes | ptr-miR-744 | MIMAT0008317 |
| View More Rows | ||
| Pig | ssc-miR-744 | MIMAT0015708 |
| Pongo pygmaeus | ppy-miR-744 | MIMAT0016102 |
| Rabbit | ocu-miR-744-5p | MIMAT0048563 |
Stem-loop Details
Human
Mouse
Bovine
Dasypus novemcinctus
Guinea pig
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit

Genomic Map