Assay Details
Gene Family ID: | MIPF0000431, mir-744 |
Mature miRNA Sequence: | UGCGGGGCUAGGGCUAACAGCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-744-5p | MIMAT0004945 |
Mouse | mmu-miR-744-5p | MIMAT0004187 |
Bovine | bta-miR-744 | MIMAT0009369 |
Dasypus novemcinctus | dno-miR-744-5p | MIMAT0048008 |
Guinea pig | cpo-miR-744-5p | MIMAT0047379 |
Pan troglodytes | ptr-miR-744 | MIMAT0008317 |
View More Rows | ||
Pig | ssc-miR-744 | MIMAT0015708 |
Pongo pygmaeus | ppy-miR-744 | MIMAT0016102 |
Rabbit | ocu-miR-744-5p | MIMAT0048563 |
Stem-loop Details
Human
Mouse
Bovine
Dasypus novemcinctus
Guinea pig
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit

More Information
Related Products
miRBase Alias:
Human: hsa-miR-744 (v17)
Mouse: mmu-miR-744 (v17)
Mouse: mmu-miR-744 (v17)
