Assay Details
Gene Family ID: | MIPF0000139, mir-500 |
Mature miRNA Sequence: | UAAUCCUUGCUACCUGGGUGAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-500a-5p | MIMAT0004773 |
| Bovine | bta-miR-500 | MIMAT0009337 |
| Goat | chi-miR-500-5p | MIMAT0036263 |
| Horse | eca-miR-500 | MIMAT0013223 |
| Pan troglodytes | ptr-miR-500 | MIMAT0008173 |
| Pig | ssc-miR-500-5p | MIMAT0037369 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-500 | MIMAT0015922 |
| Pteropus alecto | pal-miR-500-5p | MIMAT0039949 |
| Rhesus monkey | mml-miR-500a-5p | MIMAT0006363 |
| Rhesus monkey | mml-miR-500b-5p | MIMAT0028236 |
Stem-loop Details
Human
Bovine
Goat
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

Genomic Map