Assay Details
Gene Family ID: | MIPF0000139, mir-500 |
Mature miRNA Sequence: | AAUGCACCCGGGCAAGGAUUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-501-3p | MIMAT0004774 |
| Pan troglodytes | ptr-miR-501 | MIMAT0008174 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map