Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. X - 50009516 - 50009516 [+] on 50009516 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-501-5p | MIMAT0002872 |
| Human | hsa-miR-501-3p | MIMAT0004774 |
Stem-loop Details
Human

Genomic Map