Assay Details
Gene Family ID: | MIPF0000500, mir-373 |
Mature miRNA Sequence: | GAAGUGCUUCGAUUUUGGGGUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-373-3p | MIMAT0000726 |
| Pongo pygmaeus | ppy-miR-373 | MIMAT0015861 |
| Rhesus monkey | mml-miR-373 | MIMAT0006299 |
Stem-loop Details
Human
Pongo pygmaeus
Rhesus monkey

Genomic Map