Assay Details
Gene Family ID: | MIPF0000924, mir-1228 |
Mature miRNA Sequence: | GUGGGCGGGGGCAGGUGUGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1228-5p | MIMAT0005582 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-1228-5p |
---|---|
Stem-loop Accession Number: | MI0006318 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GUGGGCGGGGGCAGGUGUGUG |
Species: | Human |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000924, mir-1228 |
Mature miRNA Sequence: | GUGGGCGGGGGCAGGUGUGUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1228-5p | MIMAT0005582 |