Assay Details
Amplicon Length: | 96 |
Chromosome Location: | Chr. 12 - 57194801 - 57194801 [+] on 57194801 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1228-3p | MIMAT0005583 |
| Human | hsa-miR-1228-5p | MIMAT0005582 |
Stem-loop Details
Human

Genomic Map