Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | GAAAGCGCUUCCCUUUGCUGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518a-3p | MIMAT0002863 |
| Pan troglodytes | ptr-miR-518a | MIMAT0008186 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map