Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 19 - 53731301 - 53731301 [+] on 53731301 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518a-5p | MIMAT0005457 |
| Human | hsa-miR-518a-3p | MIMAT0002863 |
Stem-loop Details
Human

Genomic Map