Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | GGCUACAACACAGGACCCGGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-187-5p | MIMAT0004561 |
Guinea pig | cpo-miR-187-5p | MIMAT0047058 |
Rabbit | ocu-miR-187-5p | MIMAT0048295 |
Stem-loop Details
Human
Guinea pig
Rabbit
