Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 18 - 35905118 - 35905118 [-] on 35905118 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-187-5p | MIMAT0004561 |
Human | hsa-miR-187-3p | MIMAT0000262 |
Stem-loop Details
Human
