Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UUCACAGGGAGGUGUCAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-513a-5p | MIMAT0002877 |
Pongo pygmaeus | ppy-miR-513a-5p | MIMAT0015938 |
Stem-loop Details
Human
Pongo pygmaeus
