Assay Details
Amplicon Length: | 93 |
Chromosome Location: | Chr. X - 147213911 - 147213911 [+] on 147213911 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-513a-3p | MIMAT0004777 |
| Human | hsa-miR-513a-5p | MIMAT0002877 |
Stem-loop Details
Human

Genomic Map