Assay Details
Gene Family ID: | MIPF0000558, mir-1307 |
Mature miRNA Sequence: | UCGACCGGACCUCGACCGGCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1307-5p | MIMAT0022727 |
Gorilla gorilla | ggo-miR-1307 | MIMAT0049121 |
Microcebus murinus | mmr-miR-1307 | MIMAT0049136 |
Nomascus leucogenys | nle-miR-1307 | MIMAT0049461 |
Pteropus alecto | pal-miR-1307-5p | MIMAT0040316 |
Stem-loop Details
Human
Gorilla gorilla
Microcebus murinus
Nomascus leucogenys
Pteropus alecto
