Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 10 - 103394556 - 103394556 [-] on 103394556 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1307-5p | MIMAT0022727 |
Human | hsa-miR-1307-3p | MIMAT0005951 |
Stem-loop Details
Human
