Assay Details
Gene Family ID: | MIPF0000073, mir-129 |
Mature miRNA Sequence: | AAGCCCUUACCCCAAAAAGCAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-129-2-3p | MIMAT0004605 |
Mouse | mmu-miR-129-2-3p | MIMAT0000544 |
Rat | rno-miR-129-2-3p | MIMAT0000601 |
Alligator mississippiensis | ami-miR-129b-3p | MIMAT0038139 |
Bovine | bta-miR-129-3p | MIMAT0009222 |
Chicken | gga-miR-129-1-3p | MIMAT0050057 |
View More Rows | ||
Dasypus novemcinctus | dno-miR-129-2-3p | MIMAT0047626 |
Eptesicus fuscus | efu-miR-129a | MIMAT0035001 |
Gadus morhua | gmo-miR-129-3p | MIMAT0044075 |
Goat | chi-miR-129-3p | MIMAT0035937 |
Guinea pig | cpo-miR-129-2-3p | MIMAT0046979 |
Horse | eca-miR-129b-3p | MIMAT0013046 |
Ophiophagus hannah | oha-miR-129b-3p | MIMAT0036684 |
Ophiophagus hannah | oha-miR-129b | MIMAT0036682 |
Petromyzon marinus | pma-miR-129a-3p | MIMAT0019451 |
Pig | ssc-miR-129a-3p | MIMAT0013959 |
Pongo pygmaeus | ppy-miR-129-2-3p | MIMAT0016972 |
Pteropus alecto | pal-miR-129a-3p | MIMAT0039855 |
Python bivittatus | pbv-miR-129b-3p | MIMAT0038925 |
Rabbit | ocu-miR-129-2-3p | MIMAT0048216 |
Salmo salar | ssa-miR-129-3p | MIMAT0032319 |
Taeniopygia guttata | tgu-miR-129-3p | MIMAT0014499 |
Zebrafish | dre-miR-129-3p | MIMAT0003158 |
Stem-loop Details
Human
Stem-loop ID | hsa-mir-129-2 |
Stem-loop Accession # | MI0000473 |
Stem-loop Sequence | UGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAGGGCG |
Chromosome Location | Chr. 11 - 43581394 - 43581483 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0004605
miRBase ID: hsa-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: Chr. 11 - 43581394 - 43581483 [+] on Build GRCh38 |
Stem-loop ID | mmu-mir-129-2 |
Stem-loop Accession # | MI0000585 |
Stem-loop Sequence | UGCCUUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCG |
Chromosome Location | Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000544
miRBase ID: mmu-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: Chr. 2 - 94241364 - 94241453 [-] on Build GRCm38 |
Stem-loop ID | rno-mir-129-2 |
Stem-loop Accession # | MI0000637 |
Stem-loop Sequence | AGACUGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCGC |
Chromosome Location | Chr. 3 - 83137297 - 83137393 [-] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000601
miRBase ID: rno-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: Chr. 3 - 83137297 - 83137393 [-] on Build Rnor_6.0 |
Stem-loop ID | ami-mir-129b |
Stem-loop Accession # | MI0029728 |
Stem-loop Sequence | CUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUACCUAGCCGGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038139
miRBase ID: ami-miR-129b-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | bta-mir-129-2 |
Stem-loop Accession # | MI0009729 |
Stem-loop Sequence | CUGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCAC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0009222
miRBase ID: bta-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | gga-mir-129-1 |
Stem-loop Accession # | MI0040749 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUACCUACCCGGGAAGCCCUUACCCCAAAAAGCAUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0050057
miRBase ID: gga-miR-129-1-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | dno-mir-129-2 |
Stem-loop Accession # | MI0038973 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0047626
miRBase ID: dno-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | efu-mir-129a |
Stem-loop Accession # | MI0028686 |
Stem-loop Sequence | CCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0035001
miRBase ID: efu-miR-129a Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | gmo-mir-129-1 |
Stem-loop Accession # | MI0036121 |
Stem-loop Sequence | UCUUUUUGCGGUCUGGGCUUGCUGUUCAUAACUAAUAAGCUGGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044075
miRBase ID: gmo-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | gmo-mir-129-2 |
Stem-loop Accession # | MI0036166 |
Stem-loop Sequence | UCUUUUUGCGGUCUGGGCUUGCUGUUCCCACAGCCUUAGCCAGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044075
miRBase ID: gmo-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | gmo-mir-129-3 |
Stem-loop Accession # | MI0036019 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUGAUCGUGAUCUGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044075
miRBase ID: gmo-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | gmo-mir-129-4 |
Stem-loop Accession # | MI0036109 |
Stem-loop Sequence | ACUUUUUGCGGUCUGGGCUUGCUGUCCAAUCACUAUCCAGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044075
miRBase ID: gmo-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | chi-mir-129 |
Stem-loop Accession # | MI0030612 |
Stem-loop Sequence | AAGCCGGCAGACUCUGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCACU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0035937
miRBase ID: chi-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | cpo-mir-129-2 |
Stem-loop Accession # | MI0038632 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUACAUUACUCAUAGCCGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0046979
miRBase ID: cpo-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | eca-mir-129b |
Stem-loop Accession # | MI0012789 |
Stem-loop Sequence | GCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013046
miRBase ID: eca-miR-129b-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | oha-mir-129b-1 |
Stem-loop Accession # | MI0031333 |
Stem-loop Sequence | AUGACGGGUCUUCUCUGGGUCUUUUUGCGGUCUGGGCUUUCUGGGUCUGUCCAACCCCGUAUCAGAAAGCCCUUACCCCAAAAAGCAUCUGGGGUUGUACAAUGGCAUCGCUGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036684
miRBase ID: oha-miR-129b-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | oha-mir-129b-2 |
Stem-loop Accession # | MI0031332 |
Stem-loop Sequence | ACUUUGUCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUAUAUAAGUACCUAGCCGGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAGGGCGCUUUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036682
miRBase ID: oha-miR-129b Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | pma-mir-129a |
Stem-loop Accession # | MI0017062 |
Stem-loop Sequence | UGUCUCUCGCGAAUCUUUUUGCGGUUUGGGCUUGCUGUUCAUCUAAUAGCCGGCAAGCCCUUACCCCAAAAAGCAUUCGAGGGGGAUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0019451
miRBase ID: pma-miR-129a-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | ssc-mir-129a |
Stem-loop Accession # | MI0013169 |
Stem-loop Sequence | CUUCGUGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUCGCGG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013959
miRBase ID: ssc-miR-129a-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | ppy-mir-129-2 |
Stem-loop Accession # | MI0014821 |
Stem-loop Sequence | UGCCCUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAGGGCG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0016972
miRBase ID: ppy-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | pal-mir-129a |
Stem-loop Accession # | MI0032365 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0039855
miRBase ID: pal-miR-129a-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | pbv-mir-129b |
Stem-loop Accession # | MI0030222 |
Stem-loop Sequence | GUCUUCUCUGGGUCUUUUUGCGGUCUGGGCUUUCUGGGUCUGUCCAACCCCGUAUCAGAAAGCCCUUACCCCAAAAAGCAUCCGGGGUUGUACGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038925
miRBase ID: pbv-miR-129b-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | ocu-mir-129-2 |
Stem-loop Accession # | MI0039284 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUACAUAACUCAAUAGCCGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0048216
miRBase ID: ocu-miR-129-2-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | ssa-mir-129-1 |
Stem-loop Accession # | MI0026465 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUUCAUAAAUAUCAGUCUGGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032319
miRBase ID: ssa-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | ssa-mir-129-2 |
Stem-loop Accession # | MI0026466 |
Stem-loop Sequence | CUUUUUGCGGUCUGGGCUUGCUGUUAACACACCUCAACCCAGGAAGCCCUUACCCCAAAAAGCAU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032319
miRBase ID: ssa-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | tgu-mir-129 |
Stem-loop Accession # | MI0013688 |
Stem-loop Sequence | CUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUACAUAACUACCUACCCGGGAAGCCCUUACCCCAAAAAGCAUUCGCGGAG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0014499
miRBase ID: tgu-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | dre-mir-129-2 |
Stem-loop Accession # | MI0001982 |
Stem-loop Sequence | GUCUUUCACGAAUCUUUUUGCGGUCUGGGCUUGCUGUUCUCAACUAUCAAUGGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAGGGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003158
miRBase ID: dre-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |
Stem-loop ID | dre-mir-129-4 |
Stem-loop Accession # | MI0039508 |
Stem-loop Sequence | GUCUUUCGCGAAUCUUUUUGCGGUCUGGGCUUGCUGUUCAUAAGUAUCUAGCUGGGAAGCCCUUACCCCAAAAAGCAUUUGCGGAAGGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003158
miRBase ID: dre-miR-129-3p Mature miRNA Sequence: AAGCCCUUACCCCAAAAAGCAU Chromosome Location: NA |

More Information
Related Products










miRBase Alias:
Human: hsa-miR-129-3p (v17)
Mouse: mmu-miR-129-3p (v15)
Rat: rno-miR-129-2* (v18)
Rat: rno-miR-129* (v15)
Danio rerio: dre-miR-129* (v18)
Phocea massiliensis: pma-miR-129a* (v18)
Pongo pygmaeus: ppy-miR-129-2* (v18)
Staphylococcus schleiferi: ssc-miR-129 (v16)
Staphylococcus schleiferi: ssc-miR-129a (v20)
Trabulsiella guamensis ATCC 49490: tgu-miR-129 (v18)
Mouse: mmu-miR-129-3p (v15)
Rat: rno-miR-129-2* (v18)
Rat: rno-miR-129* (v15)
Danio rerio: dre-miR-129* (v18)
Phocea massiliensis: pma-miR-129a* (v18)
Pongo pygmaeus: ppy-miR-129-2* (v18)
Staphylococcus schleiferi: ssc-miR-129 (v16)
Staphylococcus schleiferi: ssc-miR-129a (v20)
Trabulsiella guamensis ATCC 49490: tgu-miR-129 (v18)
