Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 3 - 83136949 - 83136949 [-] on 83136949 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-129-2-3p | MIMAT0000601 |
| Rat | rno-miR-129-5p | MIMAT0000600 |
Stem-loop Details
Rat

Genomic Map