Assay Details
Gene Family ID: | MIPF0000139, mir-500 |
Mature miRNA Sequence: | AAUCCUUUGUCCCUGGGUGAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-501-5p | MIMAT0002872 |
| Rhesus monkey | mml-miR-501-5p | MIMAT0006364 |
Stem-loop Details
Human
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-501 (v9.2)
Macaca mulatta: mml-miR-501 (v19)
Macaca mulatta: mml-miR-501 (v19)

Genomic Map