Assay Details
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | CAGCAGCAAUUCAUGUUUUGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-322-5p | MIMAT0000548 |
| Rat | rno-miR-322-5p | MIMAT0001619 |
Stem-loop Details
Mouse
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-322 (v17)
Mouse: mmu-miR-424 (v9.2)
Rat: rno-miR-424 (v9.2)
Rat: rno-miR-322 (v18)
Mouse: mmu-miR-424 (v9.2)
Rat: rno-miR-424 (v9.2)
Rat: rno-miR-322 (v18)

Genomic Map