Assay Details
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | CAGCAGCAAUUCAUGUUUUGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-322-5p | MIMAT0000548 |
Rat | rno-miR-322-5p | MIMAT0001619 |
Stem-loop Details
Mouse
Rat
