Assay Details
Gene Family ID: | MIPF0000418, mir-590 |
Mature miRNA Sequence: | GAGCUUAUUCAUAAAAGUGCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-590-5p | MIMAT0003258 |
| Mouse | mmu-miR-590-5p | MIMAT0004895 |
| Gorilla gorilla | ggo-miR-590 | MIMAT0024206 |
| Pongo pygmaeus | ppy-miR-590-5p | MIMAT0016032 |
| Rhesus monkey | mml-miR-590-5p | MIMAT0006450 |
Stem-loop Details
Human
Mouse
Gorilla gorilla
Pongo pygmaeus
Rhesus monkey

Genomic Map