Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 7 - 74191496 - 74191496 [-] on 74191496 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-590-5p | MIMAT0003258 |
| Human | hsa-miR-590-3p | MIMAT0004801 |
Stem-loop Details
Human

Genomic Map