Assay Details
Gene Family ID: | MIPF0000393, mir-598 |
Mature miRNA Sequence: | UACGUCAUCGUUGUCAUCGUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-598-3p | MIMAT0003266 |
Gorilla gorilla | ggo-miR-598 | MIMAT0024089 |
Horse | eca-miR-598 | MIMAT0012922 |
Pan troglodytes | ptr-miR-598 | MIMAT0008267 |
Pongo pygmaeus | ppy-miR-598 | MIMAT0016040 |
Rhesus monkey | mml-miR-598-3p | MIMAT0006455 |
Stem-loop Details
Human
Gorilla gorilla
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-598 (v19)
Macaca mulatta: mml-miR-598 (v19)
Macaca mulatta: mml-miR-598 (v19)
