Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 8 - 11034906 - 11034906 [-] on 11034906 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-598-5p | MIMAT0026620 |
| Human | hsa-miR-598-3p | MIMAT0003266 |
Stem-loop Details
Human

Genomic Map