Assay Details
Gene Family ID: | MIPF0000318, mir-488 |
Mature miRNA Sequence: | CCCAGAUAAUAGCACUCUCAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-488-5p | MIMAT0003449 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-488 (v9.2)
Mouse: mmu-miR-488* (v17)
Mouse: mmu-miR-488* (v17)

Genomic Map