Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 1 - 158505930 - 158505930 [+] on 158505930 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-488-5p | MIMAT0003449 |
| Mouse | mmu-miR-488-3p | MIMAT0003450 |
Stem-loop Details
Mouse

Genomic Map