Assay Details
Gene Family ID: | MIPF0000534, mir-625 |
Mature miRNA Sequence: | AGGGGGAAAGUUCUAUAGUCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-625-5p | MIMAT0003294 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-625-5p |
---|---|
miRBase Accession Number: | MI0003639 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGGGGAAAGUUCUAUAGUCC |
Species: | Human |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000534, mir-625 |
Mature miRNA Sequence: | AGGGGGAAAGUUCUAUAGUCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-625-5p | MIMAT0003294 |