Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 14 - 65470906 - 65470906 [-] on 65470906 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-625-3p | MIMAT0004808 |
Human | hsa-miR-625-5p | MIMAT0003294 |
Stem-loop Details
Human
