Assay Details
Gene Family ID: | MIPF0000167, mir-370 |
Mature miRNA Sequence: | GCCUGCUGGGGUGGAACCUGGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-370-3p | MIMAT0000722 |
Mouse | mmu-miR-370-3p | MIMAT0001095 |
Rat | rno-miR-370-3p | MIMAT0003122 |
Bovine | bta-miR-370 | MIMAT0009300 |
Dog | cfa-miR-370 | MIMAT0009889 |
Eptesicus fuscus | efu-miR-370 | MIMAT0034931 |
View More Rows | ||
Guinea pig | cpo-miR-370-3p | MIMAT0047202 |
Horse | eca-miR-370 | MIMAT0013142 |
Pan paniscus | ppa-miR-370 | MIMAT0049048 |
Pan troglodytes | ptr-miR-370 | MIMAT0008120 |
Pig | ssc-miR-370 | MIMAT0025373 |
Pongo pygmaeus | ppy-miR-370 | MIMAT0015857 |
Rabbit | ocu-miR-370-3p | MIMAT0048405 |
Rhesus monkey | mml-miR-370-3p | MIMAT0006295 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Eptesicus fuscus
Guinea pig
Horse
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-370 (v19)
Mouse: mmu-miR-370 (v17)
Rat: rno-miR-370 (v18)
Macaca mulatta: mml-miR-370 (v19)
Mouse: mmu-miR-370 (v17)
Rat: rno-miR-370 (v18)
Macaca mulatta: mml-miR-370 (v19)
