Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUACAAAGGGAAGCCCUUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520d-5p | MIMAT0002855 |
| Pongo pygmaeus | ppy-miR-520d-5p | MIMAT0015971 |
| Pongo pygmaeus | ppy-miR-520i | MIMAT0036452 |
Stem-loop Details
Human
Pongo pygmaeus
More Information
Related Products
miRBase Alias:
Macaca mulatta: mml-miR-520d-5p (v15)
Macaca mulatta: mml-miR-518a-5p (v19)
Macaca mulatta: mml-miR-518a-5p (v19)

Genomic Map