Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAACUGUAAUUACUUUUGUAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548g-3p | MIMAT0005912 |
| Callithrix jacchus | cja-miR-548q | MIMAT0039575 |
Stem-loop Details
Human
Callithrix jacchus

Genomic Map