Assay Details
Amplicon Length: | 150 |
Chromosome Location: | Chr. 4 - 147344422 - 147344422 [+] on 147344422 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548g-3p | MIMAT0005912 |
| Human | hsa-miR-548g-5p | MIMAT0022722 |
Stem-loop Details
Human

Genomic Map