Assay Details
Mature miRNA Sequence: | GCGCAUGUUCUAUGGUCAACCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3579 | MIMAT0017867 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-3579 |
|---|---|
| miRBase Accession Number: | MI0015452 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCGCAUGUUCUAUGGUCAACCA |
| Species: | Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | GCGCAUGUUCUAUGGUCAACCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3579 | MIMAT0017867 |